Robotic tips for FLIPR Liberty 384 (Type B)
Supplier: Thermo Scientific
737-2017EA 1260 EUR
737-2017
Robotic tips for FLIPR Liberty 384 (Type B)
Pipette Tips
High quality tips, especially intended for research in molecular biology laboratories. Certified free of RNase, DNase, DNA and pyrogens.
  • Transparent PP, autoclavable
  • Very low coefficient of variation
  • Tips with ART® filters eliminate any risk of cross-contamination between samples; designed for applications with DNA, RNA as well as radioactive and infectious samples
  • Sterile tips: Sterilised by gamma radiation