Hybridisation incubator/shaker, Model S130H
Supplier: Cole Parmer
390-0243EA 5080 EUR
STUASI30H
Hybridisation incubator/shaker, Model S130H
Versatile and compact 20 litre hybridisation incubator and combined rocker shaker. The incubator temperature is accurately set via easy-to-use encoder controls and displayed by a bright LED. Forced air circulation combined with microprocessor control means outstanding temperature uniformity. The ‘safety first' design includes an over-temperature protection system that automatically tracks the set temperature and controls the heater in the event of a fault. The smart system displays error codes indicating a fault has occurred or if the set temperature has been interrupted. The incubator also cuts power to the motor if it overheats or stalls. The incubator has a stainless steel interior and a double glased, smoked acrylic and polycarbonate door which provides radiation protection. The ‘up and over' door mechanism gives excellent access to the incubator. The unit can be converted to a rocker shaker simply by removing the rotisserie and pulling forward the platform located at the rear of the incubator. The gentle rocking action is ideal for the washing stage of most hybridisation procedures. Rocking speed is fully variable allowing optimisation depending on application. Hybridisation bottles are available made from tough Pyrex® glass and leak-proof PTFE faced screw caps.
  • Rotisserie and rocker in one unit
  • Up and over door for accessibility
  • Accurate temperature control and variable speed control
  • Advanced safety features
  • BioCote® antimicrobial protection
IP Protection Class according to DIN EN 60529: IP 30