Copper ≥99.999% (metals basis), powder, Puratronic® -22 mesh

Supplier: Thermo Scientific
Warning

10609.30EA 0 EUR
10609.30
Copper ≥99.999% (metals basis), powder, Puratronic® -22 mesh
Copper
Formula: Cu
MW: 63,55 g/mol
Boiling Pt: 2595 °C (1013 hPa)
Melting Pt: 1083 °C
Density: 8,94 g/cm³ (20 °C)
MDL Number: MFCD00010965
CAS Number: 7440-50-8
ADR: 9,III
Merck Index: 13,02545

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR