Copper, gauze woven from wire 60 mesh, Ø 0.19 mm (0.0075 in)
45142.HBEA
56.2
EUR
45142.HB
45142.GT
Copper, gauze woven from wire 60 mesh, Ø 0.19 mm (0.0075 in)
Copper
Formula:
Cu MW: 63,55 g/mol Boiling Pt: 2595 °C (1013 hPa) Melting Pt: 1083 °C Density: 8,94 g/cm³ (20 °C) |
MDL Number:
MFCD00010965 CAS Number: 7440-50-8 Merck Index: 13,02545 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...