Zinc 97,5% (metals basis), powder 4 - 7 µm

Supplier: Thermo Scientific

10835.D9EA 273 EUR
10835.D9 10835.A1 10835.22
Zinc 97,5% (metals basis), powder 4 - 7 µm
Zinc
Formula: Zn
MW: 65,39 g/mol
Boiling Pt: 907 °C (1013 hPa)
Melting Pt: 419,5 °C
Density: 7,14
MDL Number: MFCD00011291
CAS Number: 7440-66-6
UN: 1436
ADR: 4.3,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR