96-well imaging plates, Falcon®
Supplier: Corning Life Sciences
Falcon®
734-2480EA 323.07 EUR
734-2480
96-well imaging plates, Falcon®
Microplates Cell Culture Plates Standard Plates
PS microplate with tissue culture treated surface.
  • Flat bottom, with lid
  • Black wells with clear bottom
  • Working volume: 25 to 340 µl