Reload systems and starter kits, DeckWorks™
Supplier: Corning Life Sciences
Corning® DeckWorks™
613-1519EA 265.23 EUR
613-1519 613-1518 613-1509 613-1516 613-1524 613-1525 613-1521 613-1507 613-1522 613-1510 613-1517 613-1523 613-1508
Reload systems and starter kits, DeckWorks™
Pipette Tips
These reload systems consist of reload decks packaged in transparent trays and filled with tips. They can be used for convenient and economical reloading of the durable DeckWorks™ hinged racks. The pipette tips are made from virgin medical grade resins, the reload decks and rack bases are manufactured from 100% recycled PP.
  • Reload decks are packaged in transparent trays for easy tip identification
  • Require no transfer devices, save space and minimise packaging waste
  • Non sterile decks and racks are autoclavable
The low binding tips feature a reduced surface tension on the interior wall of the tip. Leaching and/or subsequent sample degradation associated with  silicone-based tips are eliminated. The low binding tips are ideal for quantitative assays in which sample binding can significantly alter results. Specific applications include accurate and precise pipetting of expensive reagents, DNA, proteins and/or peptides.
Ordering information: The starter kits contain one DeckWorks™ reload system with standard tips and empty hinged racks.