Acetic acid 100%, BioChemica
Supplier: PanReac AppliChem
A3701.2500PEEA
90.7
EUR
A3701.2500PE
A3701.1000PE
Acetic acid 100%, BioChemica
Acetic acid
Formula:
H₃CCOOH MW: 60,05 g/mol Boiling Pt: 118 °C (1013 hPa) Melting Pt: 17 °C Density: 1,05 g/cm³ (20 °C) Flash Pt: 38,5 °C |
MDL Number:
MFCD00036152 CAS Number: 64-19-7 UN: 2789 ADR: 8,II |
![]() |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...