2-Chloropropane, Supelco®
Supplier: Merck
M-Clarity™ quality level = MQ100
02489-5MLEA
144
EUR
02489-5ML
2-Chloropropane, Supelco®
2-Chloropropane
Formula:
C₃H₇Cl MW: 78,54 g/mol Boiling Pt: 35,74 °C (1013 hPa) Melting Pt: –117 °C Density: 0,86 g/cm³ (20 °C) Flash Pt: –42 °C |
MDL Number:
MFCD00000867 CAS Number: 75-29-6 UN: 2356 ADR: 3,I Merck Index: 13,05231 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...