Trifluoroacetic acid, Supelco®
Supplier: Merck
M-Clarity™ quality level = MQ100
74564-1ML-FEA
31.8
EUR
74564-1ML-F
74564-10ML-F
74564-5ML-F
Trifluoroacetic acid, Supelco®
Trifluoroacetic acid
Formula:
C₂HF₃O₂ MW: 114,02 g/mol Boiling Pt: 71,8 °C (1013 hPa) Melting Pt: –15 °C Density: 1,490 g/cm³ (20 °C) |
MDL Number:
MFCD00004169 CAS Number: 76-05-1 UN: 2699 ADR: 8,I Merck Index: 12,09812 |
![]() |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...