Nitromethane, Supelco®


M-Clarity™ quality level = MQ100

02484-1MLEA 48 EUR
02484-1ML
Nitromethane, Supelco®
Nitromethane
Formula: CH₃NO₂
MW: 61,04 g/mol
Boiling Pt: 101,2 °C (1013 hPa)
Melting Pt: –29 °C
Density: 1,138 g/cm³ (20 °C)
Flash Pt: 44 °C
MDL Number: MFCD00007400
CAS Number: 75-52-5
UN: 1261
ADR: 3,II
Merck Index: 12,06708

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR