Glycerine ≥99% for biotechnology

0854-4LEA 303 EUR
0854-4L 0854-19L 0854-EPI-207L
Glycerine ≥99% for biotechnology
Glycerine

Used as a fixative in bacterial preservation media.

Formula: HOCH₂CH(OH)CH₂OH
MW: 92,09 g/mol
Boiling Pt: 290 °C (1013 hPa)
Melting Pt: 18,6 °C
Density: 1,26 g/cm³ (20 °C)
Flash Pt: 177 °C (open cup)
MDL Number: MFCD00004722
CAS Number: 56-81-5
Merck Index: 13,04497
https://export.vwr.com/stibo/web/std.lang.all/47/74/734774.jpg

Order Now

Specification Test Results

Arsenic <0.0003%
Chloride ≤0.001%
DNase none detected
Heavy metals (as Pb) <0.0005 %
Protease none detected
Purity (anhydrous) >99%
Residue on ignition <0.01%
RNase none detected
Specific Gravity Measurement 1.2615 – 1.2635
Sulphate <0.001%
Water (KF) <1%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR