Multidishes, Nunc™ Nunclon Vita
Supplier: Thermo Scientific
Nunc™ Nunclon Vita
NUNC145380EA 33.7 EUR
NUNC145380
Multidishes, Nunc™ Nunclon Vita
Microplates Cell Culture Plates Standard Plates
PS, flat bottomed, treated surface.
  • Animal component-free surface for growth of human stem cells
  • No coating or feeder cells necessary
  • Supports attachment and colony formation of human ESC and human IPS
  • Supports growth of pluripotent human ESC for at least 10 passages
  • Non-enzymatic release and dissociation with removal of ROCK inhibitor