DL-α-Tocopherol

Supplier: Enzo Life Sciences

ALEXALX460018G001EA 64.4 EUR
ALEXALX460018G001
DL-α-Tocopherol
DL-α-Tocopherol (Vitamin E)
Formula: C₂₉H₅₀O₂
MW: 430,71 g/mol
MDL Number: MFCD00072051
CAS Number: 10191-41-0
https://export.vwr.com/stibo/web/std.lang.all/07/51/45390751.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR