DL-α-Tocopherol, Sigma-Aldrich®
Supplier: Merck
M-Clarity™ quality level = MQ100
SIALT3251-100GEA
181
EUR
SIALT3251-100G
SIALT3251-500G
SIALT3251-5G
DL-α-Tocopherol, Sigma-Aldrich®
DL-α-Tocopherol (Vitamin E)
Formula:
C₂₉H₅₀O₂ MW: 430,71 g/mol |
MDL Number:
MFCD00072051 CAS Number: 10191-41-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...