n-Undecane, Supelco®
Supplier: Merck
M-Clarity™ quality level = MQ100
94000-25MLEA
211
EUR
94000-25ML
n-Undecane, Supelco®
n-Undecane
Formula:
H₃C(CH₂)₉CH₃ MW: 156,31 g/mol Boiling Pt: 196 °C (1013 hPa) Melting Pt: –26,7 °C Density: 0,7416 g/cm³ (20 °C) Flash Pt: 60 °C |
MDL Number:
MFCD00008959 UN: 2330 ADR: 3,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...