Membrane filters, track etched for cell culture applications, Whatman™
Supplier: Whatman products (Cytiva)
Whatman™
515-2291EA 171 EUR
515-2291
Membrane filters, track etched for cell culture applications, Whatman™
Filters Membrane Filters
These cell culture membrane filters are made with PVP-free PC for enhanced cellular adhesion in biological applications such as neutrophil chemotaxis.
  • For the analysis of cell migration toward a chemical stimulus
  • Thin and uniform; cylindrical pores facilitate rapid cell migration
  • Reduces incubation time and the need to sterilise
  • Offered without the standard wetting agent (PVP free membranes) for increased cellular adhesion (e.g., neutrophil chemotaxis)