Diethylnitrosoamine

442687.EA 242 EUR
442687.
Diethylnitrosoamine
Diethylnitrosoamine
Formula: C₄H₁₀N₂O
MW: 102,14 g/mol
MDL Number: MFCD00013890
CAS Number: 55-18-5
UN: 2810
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR