Serotonin hydrochloride

Supplier: Enzo Life Sciences

ALEXALX550328M250EA 161 EUR
ALEXALX550328M250 ALEXALX550328M050 ALEXALX550328G001
Serotonin hydrochloride
Serotonin hydrochloride
Formula: C₁₀H₁₃ClN₂O
MDL Number: MFCD00012686
UN: 2811
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR