Bis(2-ethylhexyl)adipate, Sigma-Aldrich®

Supplier: Merck


M-Clarity™ quality level = MQ100

SIAL02140-1LEA 23.8 EUR
SIAL02140-1L
Bis(2-ethylhexyl)adipate, Sigma-Aldrich®
Bis(2-ethylhexyl)adipate
Formula: C₂₂H₄₂O₄
MW: 370,57 g/mol
MDL Number: MFCD00009496
CAS Number: 103-23-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR