Bis(2-ethylhexyl) adipate

Supplier: Merck

442492.EA 41.1 EUR
442492.
Bis(2-ethylhexyl) adipate
Bis(2-ethylhexyl)adipate
Formula: C₂₂H₄₂O₄
MW: 370,57 g/mol
MDL Number: MFCD00009496
CAS Number: 103-23-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR