Cesium chloride ≥99.9%, Ultra Pure Grade
|
Warning
|
0415-250GEA
294
EUR
0415-250G
0415-50G
0415-100G
Cesium chloride ≥99.9%, Ultra Pure Grade
Cesium chloride
A high purity reagent used to prepare density gradients for isolation of plasmid DNA, phage or cellular RNA.
Specification Test Results
Abs.@260 nm (50%, Water) | ≤0.05 |
Chromium | ≤0.0001% |
Copper | ≤0.0001% |
Iron | ≤0.0001% |
Lead | ≤0.0001% |
Lithium | <0.001% |
Magnesium | <0.0001% |
Nickel | <0.0001% |
Potassium | <0.001% |
Purity | >99.9% |
Rubidium | <0.03% |
Sodium | <0.03% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...