Potassium chloride ≥99.0%
Supplier: Molekula
MOLE16227139-500GEA
0
EUR
MOLE16227139-500G
Potassium chloride ≥99.0%
Potassium chloride
Formula:
KCl MW: 74,55 g/mol Boiling Pt: 1420 °C (1013 hPa) Melting Pt: 773 °C Density: 1,98 g/cm³ (20 °C) |
MDL Number:
MFCD00011360 CAS Number: 7447-40-7 Merck Index: 13,07704 |
Specification Test Results
Physical appearance | White crystals or powder |
As | <0.0002% |
Assay | 99.0% min. |
Loss on drying | 1% max. |
Heavy metals | <0.0005% |
Sodium | <0.5% |
Lead | <2 mg/kg |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...