Water, Ultrapure for spectrophotometry

Supplier: Thermo Scientific

19391.M1EA 177 EUR
19391.M1 19391.K2
Water, Ultrapure for spectrophotometry
Water
Formula: H₂O
MW: 18,02 g/mol
Boiling Pt: 100 °C
Melting Pt: 0 °C
Density: 1,000
MDL Number: MFCD00011332
CAS Number: 7732-18-5
Merck Index: 13,10098

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR