Sodium dithionite ≥85%, TECHNICAL

Supplier: VWR Chemicals

27888.293EA 50.9 EUR
27888.293
Sodium dithionite ≥85%, TECHNICAL
Sodium dithionite
Formula: Na₂S₂O₄
MW: 174,11 g/mol
Melting Pt: 100 °C
Density: 2,19 g/cm³ (20 °C)
Flash Pt: 101 °C
MDL Number: MFCD00011640
UN: 1384
ADR: 4.2,II
REACH: 01-2119520510-57
Merck Index: 12,08771

Order Now

Specification Test Results

Assay Min. 85%
Identification Passes test

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR