Buffer strips and gradients, PhastGel™
Supplier: Cytiva
17-0517-01EA 102.53 EUR
17-0517-01 17-0678-01 17-0516-01 17-0542-01
Buffer strips and gradients, PhastGel™
Buffers Electrophoresis Buffers Running Buffers
PhastGel™ buffer strips are supplied as ready-to-use agarose blocks, making buffer preparation unnecessary. PhastGel™ buffer strips are specifically tested for DNA separations. PhastGel™ Gradient 8 to 25, 4 to 15, and 10 to 15 have continuous acrylamide gradients in the ranges 8% to 25%, 4% to 15% and 10% to 15%, respectively.