Ficoll-Paque™ PLUS
Supplier: Cytiva
Ficoll-Paque™
17-1440-03EA 734 EUR
17-1440-03 17-1440-02
Ficoll-Paque™ PLUS
Cell Separation Media
Ficoll-Paque™ PLUS is a sterile density gradient centrifugation medium for separation of mononuclear cells from human blood.
  • Maintains viability and representative distribution of B and T lymphocytes
  • Low levels of endotoxin (<0,12 EU/ml)
Sterile, ready to use reagents used to prepare mononuclear cell preparations from peripheral blood, bone marrow, and umbilical cord blood by density gradient centrifugation. Liquid physical state. Colorless to slightly yellow. Manufactured under a Quality Management System certified to ISO 13485:2003. Ficoll-Paque® reagents are designed to prepare human mononuclear cells from peripheral blood, bone marrow, and umbilical cord blood via density gradient centrifugation. These sterile density media have established a proven record of isolating high yields of mononuclear cells.