These ambidextrous gloves with beaded cuffs are made of 100% natural rubber latex. Thanks to their tensile strength and elasticity, they provide an excellent barrier performance and enhanced protection.
- Non sterile, powder-free
- Textured surface provides excellent grip
- Latex provides exceptional flexibility and comfort
- Beaded cuff provides easier donning and stability on the arm
- Protein level <50 µg/gram
Length: 300 mm
EN 455 (Parts 1, 2 & 3); AQL 1,5
Packaging: Packaging: 100 pieces per dispenser box; 10 dispenser boxes per carton (1000 pieces).
Certifications: EN 374-1 (Type C), EN 374-5 VIRUS, EN 421:2010; Cat. III; AQL 1,5
Food contact approved