Solar-powered thermometers, Traceable, VWR®
620-2041EA 43.5 EUR
VWRI620-2041
Solar-powered thermometers, Traceable, VWR®
This thermometer is always on, and runs for years powered by light in laboratories or plant. It can monitor ambient temperature or temperatures of liquids, air/gas and semi-solids. Perfect thermometer for measuring temperatures of solutions in tanks, water baths, incubators and refrigerators. The external probe is designed for food inspection, petroleum tests, quality control, biomedical studies, R&D analysis and wastewater tests.
  • Large LCD easily readable from across the room
  • Sensor and cable perform accurately even when under water
  • Display updates every 10 seconds to save time and eliminate 3-minute lag inherent in glass and dial thermometers
  • Reads in °F or °C at the press of a button
Internal sensor/external probe sensor (Ø: 4,7 mm) with 1 metre cable.
Certifications: Traceable to NIST for accuracy. Multi-point calibration on an individually numbered Traceable® certificate which ensures accuracy from an ISO/IEC 17025:2005 (1750.01) calibration laboratory accredited by A2LA. It indicates traceability of measurements to the SI units through NIST or other recognised national measurement institutes (NMI) that are signatories to the CIPM Mutual Recognition Agreement.
Delivery information: Supplied with solar panels, back-up battery for dim areas, VELCRO® to attach to any surface and Traceable certificate of calibration.