Energy regulator for boiling baths
Supplier: Nickel-Electro
452-0013EA 324 EUR
452-0013
Energy regulator for boiling baths
Power Supplies
This compact energy regulator is ideal for use with electric round and rectangular boiling baths excluding the 110-ER models. Suitable for 230 V, 50 to 60 Hz operation up to 10 amps.
  • Allows the boiling to be controlled from vigorous to gentle simmer
Ordering information: Supplied with mains inlet and outlet leads.