Triazophos (O,O-diethyl-O-1-phenyl-1H-1,2,4-triazol-3-yl phosphorothioate) ≥70% in xylene, Supelco®
45696-50MGEA
87.6
EUR
45696-50MG
Triazophos (O,O-diethyl-O-1-phenyl-1H-1,2,4-triazol-3-yl phosphorothioate) ≥70% in xylene, Supelco®
Triazophos (O,O-diethyl-O-1-phenyl-1H-1,2,4-triazol-3-yl phosphorothioate)
Formula:
C₁₂H₁₆N₃O₃PS MW: 313,32 g/mol |
CAS Number:
24017-47-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...