Glycylglycine (Gly-Gly)

Supplier: Merck
Sigma-Aldrich®
SIALG3915-500GEA 1030 EUR
SIALG3915-500G SIALG3915-1KG SIALG3915-25G SIALG3915-100G
Glycylglycine (Gly-Gly)
Proteins and Peptides
  • EINECS: 209-127-8
  • GHS symbol and signal word: GHS07 and warning
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR