Zinc acetate dihydrate 98-102% USP

E982-500GEA 293 EUR
E982-500G
Zinc acetate dihydrate 98-102% USP
Zinc acetate dihydrate
Formula: (H₃CCOO)₂Zn·2H₂O
MW: 219,51 g/mol
Melting Pt: 237 °C
Density: 1,718 g/cm³ (20 °C)
MDL Number: MFCD00066961
CAS Number: 5970-45-6
UN: 3077
ADR: 9,III
Merck Index: 12,10256

Order Now

Specification Test Results

Alkalies & Alkaline Earths <0.2%
Arsenic <3 ppm
Chloride <0.005 %
Insolubles <0.005 %
Lead <0.002 %
pH (5%, Water) @25°C 6 - 8
Purity 98 - 102 %
Sulphate <0.01 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR