Water cell culture reagent, Milli-Q®

Supplier: Merck


730-1371EA 0 EUR
730-1371
Water cell culture reagent, Milli-Q®
Water

Water for cell culture has been developed for laboratories requiring less than 50 litres of water/year. Intended for research use only, it has been tested and is suitable for use in experiments requiring sterile, mycoplasma-free and pyrogen-free water.


  • Sterile and free of pyrogens, mycoplasma, calcium and magnesium
  • Manufactured by a trusted leader in water purification and cell culture products and techniques
  • One label can be peeled off and affixed to a laboratory notebook for traceability in experiments
  • Available in a variety of pack sizes to suit user needs
  • Cell culture tested


Produced in an ISO® 9001 / cGMP environment. Delivered with a Certificate of Quality.

Formula: H₂O
MW: 18,02 g/mol
MDL Number: MFCD00011332
CAS Number: 7732-18-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR