Mini submarine gel electrophoresis unit, SUB6
Supplier: Hoefer
732-2076EA 450 EUR
HOEFSUB6
Mini submarine gel electrophoresis unit, SUB6
For routine preparatory and analytical electrophoresis techniques, the SUB6 mini horizontal gel unit has a maximum 32-sample throughput capacity and features a removable gel casting tray with end gaskets that allow gels to be cast directly within the tank.
  • Colour coded and height adjustable combs allow identification of comb thickness at a glance and control of well depth
  • Coloured loading strips for easy well detection when loading
  • Compact tank reduces the buffer volume required to cover the gel, providing greater control over the voltage gradient and run-time
  • UV-transparent running tray allows the user to image the gel without risk of damage due to handling
  • Side handles for safe and easy transport around the laboratory
Delivery information: System includes buffer chamber assembly, safety lid, high voltage leads, running tray with silicone gasket, 2× 1 mm thick, 8-well combs, and coloured loading strips.