Acetonitrile, J.T.Baker®

Supplier: Avantor

8149.2500EA 0 EUR
8149.2500
Acetonitrile, J.T.Baker®
Acetonitrile
Formula: H₃CCN
MW: 41,05 g/mol
Boiling Pt: 81,6 °C (1013 hPa)
Melting Pt: –45,7 °C
Density: 0,782 g/cm³ (20 °C)
Flash Pt: 2 °C (closed cup)
MDL Number: MFCD00001878
CAS Number: 75-05-8
UN: 1648
ADR: 3,II
Merck Index: 13,00071
https://export.vwr.com/stibo/web/std.lang.all/89/10/718910.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR