Reagent reservoirs, multi-well, Axygen®
Supplier: Corning Life Sciences
Corning® Axygen®
732-1391EA 362 EUR
732-1391 732-1390 732-1393 613-2304 732-1392 613-2307 613-2306 613-2308 AXYGRES-MW4-HP
Reagent reservoirs, multi-well, Axygen®
Reagent Reservoirs
PP, translucent.
  • High or low profile
  • Manufactured with virgin polypropylene for excellent chemical resistance
  • Trough Series: Unique "flow troughs" extending the length of the bottom surface
  • Compatible with most automation systems
  • Ideal for 8- and 12-channel applications
  • Certified RNase-/DNase-free, non pyrogenic