Reagent reservoirs, single-well, Axygen®
Supplier: Corning
Axygen®
AXYGRES-SW96LPSIEA 366 EUR
AXYGRES-SW96LPSI AXYGRESSW384HPSI AXYGRES-SW8-HP-S AXYGRESSW384LPSI AXYGRES-SW96-HP-SI 732-1395 AXYGRES-SW384-FXSI 732-1394 732-1396 732-1399 732-1398 AXYGRES-SW96HPSI 732-1401 732-1400 AXYGRES-SW12HPSI AXYGRES-SW8-HPSI AXYGRES-SW1LPSI AXYGRES-MW8-HP-SI
Reagent reservoirs, single-well, Axygen®
Reagent Reservoirs
PP, translucent.
  • Available with low, medium and high profile
  • Manufactured with virgin polypropylene for excellent chemical resistance
  • Flat bottom series: Single well, guaranteed flatness for a wide variety of applications
  • Diamond shaped well series: 96 or 384 well collection pools at the bottom surface help minimise dead volume
  • Certified RNase-/DNase-free, non pyrogenic