Pipette tips, universal
Supplier: Greiner bio-one
GREI739264_4800EA 611 EUR
GREI739264_4800 GREI739265_4800 GREI739263_4800 GREI740296_250
Pipette tips, universal
Pipette Tips
These pipette tips can be used with pipettors from a variety of different manufacturers.
  • Manufactured from high-grade PP
  • High breaking strength
  • Heat resistant up to approx. +140 °C and cold resistant down to −190 °C
  • Free of heavy metals
  • Autoclavable
Packaging: Supplied in bags (bulk), in yellow 'ST-racks' (standard racks) or in 'EL-racks' (EasyLoad® racks). EasyLoad® is a simple, time- and space-saving refill system for pipette tips. The racks consist of a PP box with attached lid and a PP insert plate. Users of the EasyLoad® refill system will find the insert plate included in the refill unit. The EasyLoad® racks are also particularly well suited for use with multi-channel pipettors, since the individual tips in the rack can be picked up with the same force.